Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6,6-Trimethyl-2,4-cycloheptadien-1-one 96.0+%, TCI America™
SDP

Catalog No. T18465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18465G Supplier TCI America Supplier No. T18465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 503-93-5
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00182537
InChI Key QNGQIURXCUHNAT-UHFFFAOYSA-N
PubChem CID 136330
ChEBI CHEBI:80820
IUPAC Name 2,6,6-trimethylcyclohepta-2,4-dien-1-one
SMILES CC1=CC=CC(CC1=O)(C)C

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
Formula Weight 150.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6,6-Trimethyl-2,4-cycloheptadien-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.