accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOHOCl

CAS RN 503000-87-1

IUPAC Name: 2-chloro-6-methoxypyridine-3-carboxylic acid
Synonyms: 2-chloro-6-methoxypyridine-3-carboxylic acid
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: JPBUYRWPJMVZKX-UHFFFAOYSA-N
SMILES: COC1=NC(Cl)=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results