Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methoxynicotinic acid, 95%, Thermo Scientific™

Catalog No. AAH34001MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34001MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH34001MD Supplier Thermo Scientific Chemicals Supplier No. H34001MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-6-methylpyridine-3-carboxylic acid (2-Chloro-6-methylnicotinic acid) may be used in the synthesis of 6-methyl-2-thiophenoxynicotinic acid, required for the preparation of europium complex for staining the nucleolus of NIH 3T3, HeLa and HDF cells.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 503000-87-1
Molecular Formula C7H6ClNO3
Quantity 250mg
MDL Number MFCD08236845
Synonym 2-chloro-6-methoxynicotinic acid, 2-chloro-6-methoxynicotinicacid, 3-pyridinecarboxylic acid, 2-chloro-6-methoxy, pubchem15182, 2-chloro-6-methoxy-pyridine-3-carboxylic acid, 2-chloro-6-methoxy-nicotinic acid, 2-chloro-6-methoxy pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-chloro-6-methoxy, 2-chloro-6-methoxy-3-pyridinecarboxylic acid
InChI Key JPBUYRWPJMVZKX-UHFFFAOYSA-N
SMILES COC1=NC(Cl)=C(C=C1)C(O)=O
IUPAC Name 2-chloro-6-methoxypyridine-3-carboxylic acid
Molecular Weight (g/mol) 187.58
PubChem CID 11769266
Formula Weight 187.6
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methoxynicotinic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.