accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50328-50-2

IUPAC Name: hydrogen N-[(1E)-3-(phenylimino)prop-1-en-1-yl]aniline chloride
Synonyms: hydrogen N-[(1E)-3-(phenylimino)prop-1-en-1-yl]aniline chloride
Molecular Weight (g/mol): 258.75
Molecular Formula: C15H15ClN2
InChi Key: PBKBURVPAHHUIK-AVUWLFEKSA-N
SMILES: [H+].[Cl-].N(\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results