Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malonaldehyde Dianilide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M128825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M128825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M128825G Supplier TCI America Supplier No. M128825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50328-50-2
Molecular Formula C15H14N2
MDL Number MFCD00054634
Synonym Malonaldehyde Bis(phenylimine) Hydrochloride

Specifications

Quantity 25 g
Formula Weight 258.75
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Malonaldehyde Dianilide Hydrochloride
EINECSNumber (3)-0386
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.