CAS RN 503473-32-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ TX-1918
A cell-permeable arylidene-cyclopentenedione derived tyrphostin that acts as a potent inhibitor for eEF2-K, while it inhibits other kinases at much higher concentrations.