Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TX-1918

Catalog No. 65520310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-520-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-520-310MG Supplier MilliporeSigma™ Supplier No. 65520310MG
Add to Cart
Add to Cart

A cell-permeable arylidene-cyclopentenedione derived tyrphostin that acts as a potent inhibitor for eEF2-K, while it inhibits other kinases at much higher concentrations.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Light Orange
Description 95% by HPLC
Target Proteins eEF2-K
CAS 503473-32-3
Molecular Formula C14H12O3
Formula Weight 228.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.