accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 503612-47-3

IUPAC Name: 1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-1H,4H,5H,6H,7H-pyrazolo[3,4-c]pyridine-3-carboxamide
Synonyms: apixaban
Molecular Weight (g/mol): 459.51
Molecular Formula: C25H25N5O4
InChi Key: QNZCBYKSOIHPEH-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1N=C(C(N)=O)C2=C1C(=O)N(CC2)C1=CC=C(C=C1)N1CCCCC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)