Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Apixaban

Catalog No. AC464495000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464495000 500 mg
AC464491000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC464495000 Supplier Thermo Scientific Chemicals Supplier No. 464495000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 503612-47-3 | C25H25N5O4 | 459.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503612-47-3
Molecular Formula C25H25N5O4
Molecular Weight (g/mol) 459.51
InChI Key QNZCBYKSOIHPEH-UHFFFAOYSA-N
IUPAC Name 1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-1H,4H,5H,6H,7H-pyrazolo[3,4-c]pyridine-3-carboxamide
SMILES COC1=CC=C(C=C1)N1N=C(C(N)=O)C2=C1C(=O)N(CC2)C1=CC=C(C=C1)N1CCCCC1=O

Specifications

Quantity 500 mg
Formula Weight 459.5
Chemical Name or Material Apixaban
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes damage to organs through prolonged or repeated exposure.

Precautionary Statement GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray.

Get medical attention/advice if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.