accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(R)SOOOOO

CAS RN 504-33-6

IUPAC Name: (2R)-2-azaniumyl-4-sulfonatobutanoate
Synonyms: (2R)-2-ammonio-4-sulfonatobutanoate
Molecular Weight (g/mol): 182.17
Molecular Formula: C4H8NO5S
InChi Key: VBOQYPQEPHKASR-GSVOUGTGSA-M
SMILES: [NH3+][C@H](CCS([O-])(=O)=O)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results