Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Homocysteic Acid 97.0+%, TCI America™
SDP

Catalog No. H01585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01585G 5 g
H01581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01585G Supplier TCI America Supplier No. H01585G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 504-33-6
Molecular Formula C4H9NO5S
Molecular Weight (g/mol) 183.178
MDL Number MFCD00007536
InChI Key VBOQYPQEPHKASR-UHFFFAOYSA-N
Synonym dl-homocysteic acid, homocysteic acid, dl-homocysteicacid, 2-amino-4-sulfobutyric acid, d,l-homocysteic acid, pubchem12443, acmc-1ap11, 2-amino-4-sulfo-butanoic acid, dl-2-amino-4-sulfobutyric acid
PubChem CID 92117
IUPAC Name 2-amino-4-sulfobutanoic acid
SMILES C(CS(=O)(=O)O)C(C(=O)O)N

Specifications

Color White
Quantity 5 g
Formula Weight 183.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Homocysteic Acid
RTECSNumber EK7713877
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.