CAS RN 504-57-4
IUPAC Name:
nonadecan-10-one
Synonyms:
10-nonadecanone
Molecular Weight (g/mol):
282.51
Molecular Formula:
C19H38O
InChi Key:
YUPOCHDBBHTUBJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC(=O)CCCCCCCCC
1
–
1
of
1
results
1
10-Nonadecanone 97.0+%, TCI America™
CAS: 504-57-4 Molecular Formula: C19H38O Molecular Weight (g/mol): 282.51 MDL Number: MFCD00009584 InChI Key: YUPOCHDBBHTUBJ-UHFFFAOYSA-N Synonym: Caprinone, Dinonyl Ketone PubChem CID: 10441 IUPAC Name: nonadecan-10-one SMILES: CCCCCCCCCC(=O)CCCCCCCCC