Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Nonadecanone 97.0+%, TCI America™
SDP

Catalog No. N06585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06585G 5 g
N065825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06585G Supplier TCI America Supplier No. N06585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 504-57-4
Molecular Formula C19H38O
Molecular Weight (g/mol) 282.51
MDL Number MFCD00009584
InChI Key YUPOCHDBBHTUBJ-UHFFFAOYSA-N
Synonym Caprinone, Dinonyl Ketone
PubChem CID 10441
IUPAC Name nonadecan-10-one
SMILES CCCCCCCCCC(=O)CCCCCCCCC

Specifications

Melting Point 58°C
Color White
Boiling Point 156°C
Quantity 5 g
Formula Weight 282.51
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10-Nonadecanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.