accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5044-24-6

CH3H3CNBr

CAS RN 5044-24-6

IUPAC Name: 1-(4-bromophenyl)-2,5-dimethyl-1H-pyrrole
Synonyms: 1-(4-bromophenyl)-2,5-dimethylpyrrole
Molecular Weight (g/mol): 250.14
Molecular Formula: C12H12BrN
InChi Key: OURHFEVMTCZQLA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results