Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)-2,5-dimethylpyrrole 98.0+%, TCI America™
SDP

Catalog No. B26365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26365G Supplier TCI America Supplier No. B26365G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5044-24-6
Molecular Formula C12H12BrN
Molecular Weight (g/mol) 250.14
MDL Number MFCD00277042
InChI Key OURHFEVMTCZQLA-UHFFFAOYSA-N
Synonym 1-4-bromophenyl-2,5-dimethyl-1h-pyrrole, 1-4-bromophenyl-2,5-dimethylpyrrole, 1-4-bromo-phenyl-2,5-dimethyl-1h-pyrrole, akos bb-3545, acmc-209kmk, bromophenyldimethylpyrrole, maybridge1_007386, 1h-pyrrole,1-4-bromophenyl-2,5-dimethyl
PubChem CID 2739609
IUPAC Name 1-(4-bromophenyl)-2,5-dimethyl-1H-pyrrole
SMILES CC1=CC=C(C)N1C1=CC=C(Br)C=C1

Specifications

Melting Point 74°C
Quantity 5 g
Formula Weight 250.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)-2,5-dimethylpyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.