accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH2P

CAS RN 5044-52-0

IUPAC Name: ethenyltriphenylphosphanium bromide
Synonyms: ethenyltriphenylphosphanium bromide
Molecular Weight (g/mol): 369.24
Molecular Formula: C20H18BrP
InChi Key: VRAYVWUMBAJVGH-UHFFFAOYSA-M
SMILES: [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)