Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylvinylphosphonium Bromide 96.0+%, TCI America™
SDP

Catalog No. T16445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16445G Supplier TCI America Supplier No. T16445G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5044-52-0
Molecular Formula C20H18BrP
Molecular Weight (g/mol) 369.24
MDL Number MFCD00011807
InChI Key VRAYVWUMBAJVGH-UHFFFAOYSA-M
Synonym vinyltriphenylphosphonium bromide, triphenyl vinyl phosphonium bromide, schweizer's reagent, ethenyltriphenylphosphonium bromide, ethenyl triphenyl phosphanium bromide, ethenyltriphenylphosphanium bromide, triphenyl-vinyl-phosphonium; bromide, pubchem16334, acmc-1aw2n, vinyltriphenylphosphine bromide
PubChem CID 2723893
IUPAC Name ethenyltriphenylphosphanium bromide
SMILES [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 5 g
Formula Weight 369.24
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylvinylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.