CAS RN 50479-11-3
IUPAC Name:
(4-ethoxy-4-oxobutyl)triphenylphosphanium bromide
Synonyms:
(4-ethoxy-4-oxobutyl)triphenylphosphanium bromide
Molecular Weight (g/mol):
457.35
Molecular Formula:
C24H26BrO2P
InChi Key:
JPZMNVPVVYVXAD-UHFFFAOYSA-M
SMILES:
[Br-].CCOC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich Copper
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.