accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 5048-30-6

IUPAC Name: hept-6-en-1-yl acetate
Synonyms: hept-6-en-1-yl acetate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: GNEUVQRNOYJQLS-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results