Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Heptenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. H08495ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08495ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H08495ML Supplier TCI America Supplier No. H08495ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5048-30-6
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00191656
InChI Key GNEUVQRNOYJQLS-UHFFFAOYSA-N
Synonym 7-Acetoxy-1-heptene, Acetic Acid 6-Heptenyl Ester
PubChem CID 537537
IUPAC Name hept-6-en-1-yl acetate
SMILES CC(=O)OCCCCCC=C

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 mL
Formula Weight 156.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Heptenyl Acetate
EINECSNumber (2)-0760
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.