accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 505-65-7

OO

CAS RN 505-65-7

IUPAC Name: 1,3-dioxepane
Synonyms: 1,3-dioxepane
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: CZLMRJZAHXYRIX-UHFFFAOYSA-N
SMILES: C1CCOCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results