accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 505-86-2

IUPAC Name: hexadecyltrimethylazanium hydroxide
Synonyms: cetrimonium hydroxide
Molecular Weight (g/mol): 301.56
Molecular Formula: C19H43NO
InChi Key: WJLUBOLDZCQZEV-UHFFFAOYSA-M
SMILES: [OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (6)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)