Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyltrimethylammonium Hydroxide (25% in Methanol), TCI America™
SDP

Catalog No. H008325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H008325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H008325ML Supplier TCI America Supplier No. H008325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 505-86-2
Molecular Formula C19H43NO
Molecular Weight (g/mol) 301.56
MDL Number MFCD00041921
InChI Key WJLUBOLDZCQZEV-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium hydroxide, cetyltrimethylammonium hydroxide, unii-8q7d6895lv, n,n,n-trimethylhexadecan-1-aminium hydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide, cetrimide inn:ban:jan, n-hexadecyltrimethylammonium hydroxide, n,n,n-trimethyl-1-hexadecanaminiuhydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide 1:1, cetyl-trimethylammonium hydroxide
PubChem CID 68166
IUPAC Name hexadecyltrimethylazanium hydroxide
SMILES [OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color Yellow
Quantity 25 mL
UN Number 2924
Formula Weight 301.56
Physical Form Liquid
Chemical Name or Material Hexadecyltrimethylammonium Hydroxide (25% in Methanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.