CAS RN 505-95-3
IUPAC Name:
12-hydroxydodecanoic acid
Synonyms:
12-hydroxydodecanoic acid ω hydroxy dodecanoate ω-OH lauric acid ω-hydroxy lauric acid ω-OH dodecanoic acid ω-hydroxydodecanoic acid
Molecular Weight (g/mol):
216.32
Molecular Formula:
C12H24O3
InChi Key:
ZDHCZVWCTKTBRY-UHFFFAOYSA-N
SMILES:
OCCCCCCCCCCCC(O)=O
1
–
2
of
2
results
1
Sigma Aldrich 12-Hydroxydodecanoic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.