Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

12-Hydroxydodecanoic Acid, ≥97%, MP Biomedicals™

Catalog No. ICN15743001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15743001 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15743001 Supplier MP Biomedicals, Inc Supplier No. 0215743001

Specifications

CAS 505-95-3
Color White
Assay Percent Range ≥97%
Molecular Formula C12H24O3
Quantity 1 g
MDL Number MFCD00002739
Synonym 12-hydroxylauric acid, 12-hydroxy-dodecanoic acid, dodecanoic acid, 12-hydroxy, 12-hydroxy lauric acid, omega-hydroxydodecanoic acid, omega-hydroxydodecanoate, sabinic acid, 12-hydroxydodecanoicacid, omega-oh lauric acid, omega hydroxy dodecanoate
InChI Key ZDHCZVWCTKTBRY-UHFFFAOYSA-N
SMILES C(CCCCCC(=O)O)CCCCCO
IUPAC Name 12-hydroxydodecanoic acid
Molecular Weight (g/mol) 216.321
PubChem CID 79034
ChEBI CHEBI:39567
Formula Weight 216.3
Physical Form Solid
Chemical Name or Material 12-Hydroxydodecanoic acid
Show More Show Less
EINECSNumber 208-025-0
Recommended Storage Store at Room Temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.