accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5053-24-7

IUPAC Name: 2-(methylsulfanyl)-1,3-thiazole
Synonyms: 2-(methylsulfanyl)-1,3-thiazole
Molecular Weight (g/mol): 131.21
Molecular Formula: C4H5NS2
InChi Key: VQNOAXZUEKPSJC-UHFFFAOYSA-N
SMILES: CSC1=NC=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results