Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)thiazole 98.0+%, TCI America™
SDP

Catalog No. M17125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17125G 5 g
M171225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17125G Supplier TCI America Supplier No. M17125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5053-24-7
Molecular Formula C4H5NS2
Molecular Weight (g/mol) 131.21
MDL Number MFCD00858981
InChI Key VQNOAXZUEKPSJC-UHFFFAOYSA-N
Synonym 2-methylthio thiazole, 2-methylthiothiazole, 2-methylthio-1,3-thiazole, 2-methylsulfanyl-1,3-thiazole, thiazole, 2-methylthio, acmc-209knn, 2-methylsulfanyl-thiazole, thiazole,2-methylthio
PubChem CID 2760100
IUPAC Name 2-(methylsulfanyl)-1,3-thiazole
SMILES CSC1=NC=CS1

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 g
Formula Weight 131.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.