accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CO(E)(E)(Z)

CAS RN 5056-17-7

IUPAC Name: (2Z,4E,6E)-2,7-dimethylocta-2,4,6-trienedial
Synonyms: (2Z,4E,6E)-2,7-dimethylocta-2,4,6-trienedial
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: AYODHZHFDRRQEZ-XDRNSHQHSA-N
SMILES: C\C(C=O)=C\C=C\C=C(/C)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results