Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2E,4E,6E)-2,7-Dimethylocta-2,4,6-trienedial 98.0+%, TCI America™
SDP

Catalog No. D45845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45845G 5 g
D458425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45845G Supplier TCI America Supplier No. D45845G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5056-17-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD11113099
InChI Key AYODHZHFDRRQEZ-XLKYRCCQSA-N
PubChem CID 6504517
IUPAC Name (2E,4E,6E)-2,7-dimethylocta-2,4,6-trienedial
SMILES CC(=CC=CC=C(C)C=O)C=O

Specifications

Melting Point 166°C
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2E,4E,6E)-2,7-Dimethylocta-2,4,6-trienedial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.