accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50593-92-5

IUPAC Name: 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Synonyms: 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Molecular Weight (g/mol): 249.08
Molecular Formula: C6H5BrN2O2S
InChi Key: YJEWVVYJOJJLBP-UHFFFAOYSA-N
SMILES: CSC1=NC=C(Br)C(=N1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results