accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 506-12-7

IUPAC Name: heptadecanoic acid
Synonyms: heptadecanoic acid margaric acid, sodium salt margaric acid, potassium salt normal-heptadecanoic acid ω i-123 heptadecanoic acid margaric acid margarinsaeure margaric acid, 1-(11)C-labeled margarinic acid 1-(11)c-labeled margaric acid
Molecular Weight (g/mol): 270.46
Molecular Formula: C17H34O2
InChi Key: KEMQGTRYUADPNZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (5)
Boiling Point 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)