Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Heptadecanoic acid, 99.1%, For GC anlysis, MP Biomedicals™

Catalog No. ICN19521605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19521605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19521605 Supplier MP Biomedicals, Inc Supplier No. 0219521605

Specifications

CAS 506-12-7
Color White
Molecular Formula C17H34O2
Quantity 5 g
Synonym margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
InChI Key KEMQGTRYUADPNZ-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCC(=O)O
IUPAC Name heptadecanoic acid
Molecular Weight (g/mol) 270.457
PubChem CID 10465
ChEBI CHEBI:32365
Formula Weight 270.5
Percent Purity 0.991
Physical Form Flaky Powder
Chemical Name or Material N-Heptadecanoic acid, For GC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.