accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH(Z)(Z)

CAS RN 506-43-4

IUPAC Name: (9Z,12Z)-octadeca-9,12-dien-1-ol
Synonyms: linoleyl alcohol
Molecular Weight (g/mol): 266.47
Molecular Formula: C18H34O
InChi Key: JXNPEDYJTDQORS-HZJYTTRNSA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results