Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis,cis-9,12-Octadecadien-1-ol 95.0+%, TCI America™
SDP

Catalog No. O02255ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02255ML 5 mL
O022525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02255ML Supplier TCI America Supplier No. O02255ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 506-43-4
Molecular Formula C18H34O
Molecular Weight (g/mol) 266.47
MDL Number MFCD00056667
InChI Key JXNPEDYJTDQORS-HZJYTTRNSA-N
Synonym Linoleyl Alcohol
PubChem CID 5365682
ChEBI CHEBI:73534
IUPAC Name (9Z,12Z)-octadeca-9,12-dien-1-ol
SMILES CCCCC\C=C/C\C=C/CCCCCCCCO

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 mL
Formula Weight 266.47
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis,cis-9,12-Octadecadien-1-ol
EINECSNumber (2)-0258
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.