CAS RN 50603-71-9
IUPAC Name:
methyl 2-oxo-2H-cyclohepta[b]furan-3-carboxylate
Synonyms:
methyl 2-oxocyclohepta[b]furan-3-carboxylate
Molecular Weight (g/mol):
204.18
Molecular Formula:
C11H8O4
InChi Key:
NWECYFMGBPWBAL-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C2C=CC=CC=C2OC1=O
1
–
1
of
1
results
1
3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one 98.0+%, TCI America™
CAS: 50603-71-9 Molecular Formula: C11H8O4 Molecular Weight (g/mol): 204.181 MDL Number: MFCD00671552 InChI Key: NWECYFMGBPWBAL-UHFFFAOYSA-N PubChem CID: 10081616 IUPAC Name: methyl 2-oxocyclohepta[b]furan-3-carboxylate SMILES: COC(=O)C1=C2C=CC=CC=C2OC1=O