Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one 98.0+%, TCI America™
SDP

Catalog No. M11881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11881G Supplier TCI America Supplier No. M11881G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50603-71-9
Molecular Formula C11H8O4
Molecular Weight (g/mol) 204.181
MDL Number MFCD00671552
InChI Key NWECYFMGBPWBAL-UHFFFAOYSA-N
PubChem CID 10081616
IUPAC Name methyl 2-oxocyclohepta[b]furan-3-carboxylate
SMILES COC(=O)C1=C2C=CC=CC=C2OC1=O

Specifications

Melting Point 173°C
Quantity 1 g
Formula Weight 204.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.