accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50606-96-7

IUPAC Name: 4-heptylbenzoyl chloride
Synonyms: 4-heptylbenzoyl chloride
Molecular Weight (g/mol): 238.76
Molecular Formula: C14H19ClO
InChi Key: WHTFLTOKFXTJGV-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C(Cl)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)