Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptylbenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. H07925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07925G 5 g
H079225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07925G Supplier TCI America Supplier No. H07925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50606-96-7
Molecular Formula C14H19ClO
Molecular Weight (g/mol) 238.755
MDL Number MFCD00000702
InChI Key WHTFLTOKFXTJGV-UHFFFAOYSA-N
Synonym 4-n-heptylbenzoyl chloride, benzoyl chloride, 4-heptyl, p-heptylbenzoyl chloride, acmc-209ko7, benzoyl chloride,4-heptyl, 4-n-heptyl-benzoyl chloride, 4-heptylbenzoyl chloride, 4-n-heptyl benzoic acid chloride, 4-hept-1-yl benzoyl chloride 95+%
PubChem CID 170864
IUPAC Name 4-heptylbenzoyl chloride
SMILES CCCCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Color Yellow
Boiling Point 196°C
Quantity 5 g
UN Number 3265
Formula Weight 238.76
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Heptylbenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.