accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 5063-65-0

IUPAC Name: 2-pentyloxirane
Synonyms: 2-pentyloxirane
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: NMOFYYYCFRVWBK-UHFFFAOYNA-N
SMILES: CCCCCC1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results