Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxyheptane 96.0+%, TCI America™
SDP

Catalog No. E03125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03125ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E03125ML Supplier TCI America Supplier No. E03125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5063-65-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00037521
InChI Key NMOFYYYCFRVWBK-UHFFFAOYNA-N
Synonym 1,2-Heptylene Oxide
PubChem CID 92215
IUPAC Name 2-pentyloxirane
SMILES CCCCCC1CO1

Specifications

Color Colorless
Boiling Point 144°C
Quantity 5 mL
UN Number 1993
Formula Weight 114.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxyheptane
EINECSNumber (2)-0220
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.