accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaOONOOOO

CAS RN 5064-31-3

IUPAC Name: trisodium 2-[bis(carboxylatomethyl)amino]acetate
Synonyms: trisodium nitrilotriacetate
Molecular Weight (g/mol): 257.08
Molecular Formula: C6H6NNa3O6
InChi Key: DZCAZXAJPZCSCU-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)