Learn More
Nitrilotriacetic Acid Trisodium Salt, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
N,N-Bis(carboxymethyl)glycine Trisodium Salt, Trisodium NTA
Specifications
Specifications
| Quantity | 1 kg |
| Molecular Weight (g/mol) | 257.09 |
| Percent Purity | 98% |
| Grade | Ungraded |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Nitrilotriacetic Acid Trisodium Salt |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.