accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3IO

CAS RN 507-02-8

IUPAC Name: acetyl iodide
Synonyms: acetyl iodide
Molecular Weight (g/mol): 169.95
Molecular Formula: C2H3IO
InChi Key: LEKJTGQWLAUGQA-UHFFFAOYSA-N
SMILES: CC(I)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results