Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl Iodide 95.0+%, TCI America™
SDP

Catalog No. A009525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A009525G 25 g
A009510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A009525G Supplier TCI America Supplier No. A009525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 507-02-8
Molecular Formula C2H3IO
Molecular Weight (g/mol) 169.949
MDL Number MFCD00039399
InChI Key LEKJTGQWLAUGQA-UHFFFAOYSA-N
PubChem CID 10483
IUPAC Name acetyl iodide
SMILES CC(=O)I

Specifications

Color Red
Boiling Point 108°C
Quantity 25 g
UN Number 1898
Formula Weight 169.95
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Acetyl Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.