accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3ClO

CAS RN 507-40-4

IUPAC Name: 2-chlorosyl-2-methylpropane
Synonyms: t-butyl hypochlorite
Molecular Weight (g/mol): 108.57
Molecular Formula: C4H9ClO
InChi Key: KNSWLJFYTHGVGP-UHFFFAOYSA-N
SMILES: CC(C)(C)[Cl]=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results