accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3OO(S)(S)(R)(S)(S)CH3(S)(S)(R)OCH3ONH3C(S)CH3(S)N

CAS RN 50700-72-6

IUPAC Name: 1-[(1R,2S,3aS,3bR,5aS,7S,8S,9aS,9bS,11aS)-1,7-bis(acetyloxy)-9a,11a-dimethyl-8-(piperidin-1-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-2-yl]-1-methylpiperidin-1-ium bromide
Synonyms: vecuronium bromide bromuro de vecuronio bromure de vecuronium vecuronii bromidum
Molecular Weight (g/mol): 637.74
Molecular Formula: C34H57BrN2O4
InChi Key: VEPSYABRBFXYIB-PWXDFCLTSA-M
SMILES: [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](OC(C)=O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N1CCCCC1)[N+]1(C)CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)