Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vecuronium Bromide Hydrate 98.0+%, TCI America™
SDP

Catalog No. V0120100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0120100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V0120100MG Supplier TCI America Supplier No. V0120100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50700-72-6
Molecular Formula C34H57BrN2O4
Molecular Weight (g/mol) 637.744
MDL Number MFCD00867762
InChI Key VEPSYABRBFXYIB-PWXDFCLTSA-M
Synonym 1-[(2beta,3alpha,5alpha,16beta,17beta)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methylpiperidinium Bromide
PubChem CID 39764
ChEBI CHEBI:9940
IUPAC Name [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-acetyloxy-10,13-dimethyl-16-(1-methylpiperidin-1-ium-1-yl)-2-piperidin-1-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;bromide
SMILES CC(=O)OC1CC2CCC3C(C2(CC1N4CCCCC4)C)CCC5(C3CC(C5OC(=O)C)[N+]6(CCCCC6)C)C.[Br-]

Specifications

Melting Point 235°C
Color White
Quantity 100 mg
Formula Weight 637.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Vecuronium Bromide Hydrate
RTECSNumber TN4875000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.