CAS RN 50729-75-4
IUPAC Name:
6-[2-(2-undecyl-1H-imidazol-1-yl)ethyl]-1,3,5-triazine-2,4-diamine
Synonyms:
6-[2-(2-undecylimidazol-1-yl)ethyl]-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol):
359.52
Molecular Formula:
C19H33N7
InChi Key:
QZDGTFKTHGKXEP-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC1=NC=CN1CCC1=NC(N)=NC(N)=N1
1
–
1
of
1
results
1
2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine 99.0+%, TCI America™
CAS: 50729-75-4 Molecular Formula: C19H33N7 Molecular Weight (g/mol): 359.522 InChI Key: QZDGTFKTHGKXEP-UHFFFAOYSA-N PubChem CID: 162605 IUPAC Name: 6-[2-(2-undecylimidazol-1-yl)ethyl]-1,3,5-triazine-2,4-diamine SMILES: CCCCCCCCCCCC1=NC=CN1CCC2=NC(=NC(=N2)N)N