Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine 99.0+%, TCI America™
SDP

Catalog No. D323925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D323925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D323925G Supplier TCI America Supplier No. D323925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50729-75-4
Molecular Formula C19H33N7
Molecular Weight (g/mol) 359.522
InChI Key QZDGTFKTHGKXEP-UHFFFAOYSA-N
PubChem CID 162605
IUPAC Name 6-[2-(2-undecylimidazol-1-yl)ethyl]-1,3,5-triazine-2,4-diamine
SMILES CCCCCCCCCCCC1=NC=CN1CCC2=NC(=NC(=N2)N)N

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 25 g
Formula Weight 359.52
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine
EINECSNumber (5)-1008
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.