accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)H3N(S)NH3CH3

CAS RN 50764-59-5

IUPAC Name: (1R,2S)-1,2-bis(4-methylphenyl)ethane-1,2-bis(aminium)
Synonyms: (1R,2S)-1,2-bis(4-methylphenyl)ethane-1,2-bis(aminium)
Molecular Weight (g/mol): 242.37
Molecular Formula: C16H22N2
InChi Key: UQUZLWQGLCLOBD-IYBDPMFKSA-P
SMILES: CC1=CC=C(C=C1)[C@@H]([NH3+])[C@@H]([NH3+])C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results